2-Methoxy-1-(4-(trifluoromethyl)phenyl)ethanone

Lieferant: Apollo Scientific

PC448024-1G
APOSPC448024-1GEA 539 EUR
APOSPC448024-1G
2-Methoxy-1-(4-(trifluoromethyl)phenyl)ethanone
2-Methoxy-1-(4-(trifluoromethyl)phenyl)ethanone
Formel: C₁₀H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09751273
CAS-Nummer: 26771-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR