Methyl-6-fluoro-1H-indole-2-carboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 6-fluoro-1H-indole-2-carboxylate

PC448025-1G
APOSPC448025-1GEA 967 EUR
APOSPC448025-1G
Methyl-6-fluoro-1H-indole-2-carboxylate
Methyl-6-fluoro-1H-indole-2-carboxylate
Formel: C₁₀H₈FNO₂
Molecular Weight: 193,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04966976
CAS-Nummer: 136818-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR