5-Methyl-3-(trifluoromethyl)-1H-pyrazole

Lieferant: Apollo Scientific

PC448028-1G
APOSPC448028-1GEA 32.2 EUR
APOSPC448028-1G
5-Methyl-3-(trifluoromethyl)-1H-pyrazole
5-Methyl-3-(trifluoromethyl)-1H-pyrazole
Formel: C₅H₅F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00156058
CAS-Nummer: 10010-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR