(S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol
Lieferant: Apollo Scientific
|
Achtung
|
PC448030-5G
APOSPC448030-5GEA
55.1
EUR
APOSPC448030-5G
(S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol
(S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol
Formel:
C₈H₇Cl₂FO Molecular Weight: 209.04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09863793 CAS-Nummer: 877397-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...