1-(2-Tetrahydropyranyl)-3-(trifluormethyl)-1H-pyrazol-5-boronsäure

Lieferant: Apollo Scientific

Synonyme: (1-(Tetrahydro-2H-pyran-2-yl)-3-(trifluoromethyl)-1H-pyrazol-5-yl)boronic acid

PC448031-250MG
APOSPC448031-250MGEA 473 EUR
APOSPC448031-250MG
1-(2-Tetrahydropyranyl)-3-(trifluormethyl)-1H-pyrazol-5-boronsäure
1-(2-Tetrahydropyranyl)-3-(trifluormethyl)-1H-pyrazol-5-boronsäure
Formel: C₉H₁₂BF₃N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD17214248
CAS-Nummer: 1141878-45-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR