2,3,5-Trifluor-4-methylbenzoesäure

Lieferant: Apollo Scientific

PC448033-1G
APOSPC448033-1GEA 326 EUR
APOSPC448033-1G
2,3,5-Trifluor-4-methylbenzoesäure
2,3,5-Trifluor-4-methylbenzoesäure
Formel: C₈H₅F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00456800
CAS-Nummer: 773869-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR