(3S,4R)-1-tert-Butyl-4-(2,4-difluorophenyl)pyrrolidine-3-carboxylic acid 98%

Lieferant: Apollo Scientific

PC448050-250MG
APOSPC448050-250MGEA 238 EUR
APOSPC448050-250MG
(3S,4R)-1-tert-Butyl-4-(2,4-difluorophenyl)pyrrolidine-3-carboxylic acid 98%
(3S,4R)-1-tert-Butyl-4-(2,4-difluorophenyl)pyrrolidine-3-carboxylic acid
Formel: C₁₅H₁₉F₂NO₂
Molecular Weight: 283,314 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13152278
CAS-Nummer: 455957-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR