tert-Butyl 2-amino-4-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific

PC448058-1G PC448058-250MG
APOSPC448058-1GEA 982 EUR
APOSPC448058-1G APOSPC448058-250MG
tert-Butyl 2-amino-4-(trifluoromethyl)benzoate
tert-Butyl 2-amino-4-(trifluoromethyl)benzoate
Formel: C₁₂H₁₄F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08274733
CAS-Nummer: 1485729-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR