3,3-Difluoro-6-methoxyindolin-2-one 95%
Lieferant: Apollo Scientific
PC448200-250MG
APOSPC448200-250MGEA
1810
EUR
APOSPC448200-250MG
3,3-Difluoro-6-methoxyindolin-2-one 95%
3,3-Difluoro-6-methoxyindolin-2-one
Formel:
C₉H₇F₂NO₂ Molecular Weight: 199,154 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22547892 CAS-Nummer: 1393560-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...