4-Bromo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine ≥98%

Lieferant: Apollo Scientific

PC448202-100MG
APOSPC448202-100MGEA 927 EUR
APOSPC448202-100MG
4-Bromo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine ≥98%
4-Bromo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₄BrF₃N₂
Molecular Weight: 265,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19443248
CAS-Nummer: 1379358-35-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR