4-[(2-Chloro-4-pyridinyl)oxy]-2,5-difluoro-benzenamine 95%
Lieferant: Apollo Scientific
PC448207-100MG
APOSPC448207-100MGEA
463
EUR
APOSPC448207-100MG
4-[(2-Chloro-4-pyridinyl)oxy]-2,5-difluoro-benzenamine 95%
4-[(2-Chloro-4-pyridinyl)oxy]-2,5-difluoro-benzenamine
Formel:
C₁₁H₇ClF₂N₂O Molecular Weight: 256,636 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26961015 CAS-Nummer: 1225278-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...