2-[(Dimethylamino)methylene]-5-(4-fluorophenyl)-1,3-cyclohexanedione 95%

Lieferant: Apollo Scientific

PC448208-500MG
APOSPC448208-500MGEA 1100 EUR
APOSPC448208-500MG
2-[(Dimethylamino)methylene]-5-(4-fluorophenyl)-1,3-cyclohexanedione 95%
2-[(Dimethylamino)methylene]-5-(4-fluorophenyl)-1,3-cyclohexanedione
Formel: C₁₅H₁₆FNO₂
Molecular Weight: 261,291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13811664
CAS-Nummer: 1030419-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR