tert-Butyl 3-(trifluoromethyl)-2,5-dihydro-1H-pyrrole-1-carboxylate 95%

Lieferant: Apollo Scientific

PC448210-100MG
APOSPC448210-100MGEA 795 EUR
APOSPC448210-100MG
tert-Butyl 3-(trifluoromethyl)-2,5-dihydro-1H-pyrrole-1-carboxylate 95%
tert-Butyl 3-(trifluoromethyl)-2,5-dihydro-1H-pyrrole-1-carboxylate
Formel: C₁₀H₁₄F₃NO₂
Molecular Weight: 237,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24468819
CAS-Nummer: 693826-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR