tert-Butyl 5-bromo-4-fluoro-5,6-dihydropyridine-1(2H)-carboxylate 95%

Lieferant: Apollo Scientific

PC448211-250MG
APOSPC448211-250MGEA 1190 EUR
APOSPC448211-250MG
tert-Butyl 5-bromo-4-fluoro-5,6-dihydropyridine-1(2H)-carboxylate 95%
tert-Butyl 5-bromo-4-fluoro-5,6-dihydropyridine-1(2H)-carboxylate
Formel: C₁₀H₁₅BrFNO₂
Molecular Weight: 280,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28975649
CAS-Nummer: 1881331-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR