tert-Butyl 4-(trifluoromethyl)-5,6-dihydropyridine-1(2H)-carboxylate 95%
Lieferant: Apollo Scientific
PC448212-100MG
APOSPC448212-100MGEA
703
EUR
APOSPC448212-100MG
tert-Butyl 4-(trifluoromethyl)-5,6-dihydropyridine-1(2H)-carboxylate 95%
tert-Butyl 4-(trifluoromethyl)-5,6-dihydropyridine-1(2H)-carboxylate
Formel:
C₁₁H₁₆F₃NO₂ Molecular Weight: 251,245 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28967983 CAS-Nummer: 291289-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...