4,4-Difluoropiperidin-3-yl acetate hydrochloride 95%

Lieferant: Apollo Scientific

PC448215-100MG
APOSPC448215-100MGEA 1050 EUR
APOSPC448215-100MG
4,4-Difluoropiperidin-3-yl acetate hydrochloride 95%
4,4-Difluoropiperidin-3-yl acetate hydrochloride
Formel: C₇H₁₁F₂NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28975627
CAS-Nummer: 1881331-81-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR