2,2,2-Trifluoro-1-(piperidin-3-yl)ethanamine dihydrochloride 95%

Lieferant: Apollo Scientific

PC448216-100MG
APOSPC448216-100MGEA 1350 EUR
APOSPC448216-100MG
2,2,2-Trifluoro-1-(piperidin-3-yl)ethanamine dihydrochloride 95%
2,2,2-Trifluoro-1-(piperidin-3-yl)ethanamine dihydrochloride
Formel: C₇H₁₃F₃N₂·2ClH
Molecular Weight: 255,109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28967968
CAS-Nummer: 1881331-20-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR