3-(3-Fluorophenoxy)-azetidine hydrochloride

Lieferant: Apollo Scientific

PC449000-1G
APOSPC449000-1GEA 319 EUR
APOSPC449000-1G
3-(3-Fluorophenoxy)-azetidine hydrochloride
3-(3-Fluorophenoxy)-azetidine hydrochloride
Formel: C₉H₁₀FNO·ClH
Molecular Weight: 203,641 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774228
CAS-Nummer: 1236861-75-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR