3-(4-Fluorophenoxy)azetidine hydrochloride

Lieferant: Apollo Scientific

PC449001-1G
APOSPC449001-1GEA 800 EUR
APOSPC449001-1G
3-(4-Fluorophenoxy)azetidine hydrochloride
3-(4-Fluorophenoxy)azetidine hydrochloride
Formel: C₉H₁₀FNO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD05664839
CAS-Nummer: 63843-78-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR