N-(Cyclopropylmethyl)-4-fluoro-benzylamine

Lieferant: Apollo Scientific

Synonyme: 1-Cyclopropyl-N-(4-fluorobenzyl)methanamine

PC449004-1G
APOSPC449004-1GEA 579 EUR
APOSPC449004-1G
N-(Cyclopropylmethyl)-4-fluoro-benzylamine
N-(Cyclopropylmethyl)-4-fluoro-benzylamine
Formel: C₁₁H₁₄FN
Molecular Weight: 179,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11140258
CAS-Nummer: 1019538-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR