N-Cyclopropyl-4-fluorobenzylamine
Lieferant: Apollo Scientific
Synonyme:
Cyclopropyl(4-fluorobenzyl)amine hydrochloride
PC449006-5G
APOSPC449006-5GEA
873
EUR
APOSPC449006-5G
N-Cyclopropyl-4-fluorobenzylamine
N-Cyclopropyl-4-fluorobenzylamine
Formel:
C₁₀H₁₂FN Molecular Weight: 165,211 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07330007 CAS-Nummer: 625437-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...