N-[4-(Trifluoromethyl)benzyl]cyclopropanamine

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Cyclopropyl-4-(trifluoromethyl)-benzylamine

PC449007-5G
APOSPC449007-5GEA 1160 EUR
APOSPC449007-5G
N-[4-(Trifluoromethyl)benzyl]cyclopropanamine
N-[4-(Trifluoromethyl)benzyl]cyclopropanamine
Formel: C₁₁H₁₂F₃N
Molecular Weight: 215,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09043527
CAS-Nummer: 643007-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR