1-[2-(3-Fluorophenyl)ethyl]piperazine dihydrochloride
Lieferant: Apollo Scientific
PC449010-1G
APOSPC449010-1GEA
771
EUR
APOSPC449010-1G
1-[2-(3-Fluorophenyl)ethyl]piperazine dihydrochloride
1-[2-(3-Fluorophenyl)ethyl]piperazine dihydrochloride
Formel:
C₁₂H₁₇FN₂·2ClH Molecular Weight: 281,197 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28975213 CAS-Nummer: 188400-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...