1-[2-(4-Fluorophenoxy)ethyl]piperazine

Lieferant: Apollo Scientific

Synonyme: 1-[2-(4-Fluorophenoxy)ethyl]piperazine dihydrochloride

PC449011-1G
APOSPC449011-1GEA 806 EUR
APOSPC449011-1G
1-[2-(4-Fluorophenoxy)ethyl]piperazine
1-[2-(4-Fluorophenoxy)ethyl]piperazine
Formel: C₁₂H₁₇FN₂O
Molecular Weight: 224,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04211513
CAS-Nummer: 77602-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR