[1-(4-Trifluoromethylphenyl)cyclopropyl]methanol

Lieferant: Apollo Scientific

PC449013-1G
APOSPC449013-1GEA 780 EUR
APOSPC449013-1G
[1-(4-Trifluoromethylphenyl)cyclopropyl]methanol
[1-(4-Trifluoromethylphenyl)cyclopropyl]methanol
Formel: C₁₁H₁₁F₃O
Molecular Weight: 216,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374457
CAS-Nummer: 850406-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR