N-Cyclopropyl-3-(trifluoromethyl)-benzylamine
Lieferant: Apollo Scientific
PC449015-1G
APOSPC449015-1GEA
960
EUR
APOSPC449015-1G
N-Cyclopropyl-3-(trifluoromethyl)-benzylamine
N-Cyclopropyl-3-(trifluoromethyl)-benzylamine
Formel:
C₁₁H₁₂F₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD08060650 CAS-Nummer: 16065-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...