1-[3-(Trifluoromethyl)phenyl]-cyclobutanecarboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 1-(3-(Trifluoromethyl)phenyl)cyclobutanecarboxylic acid

PC449018-5G
APOSPC449018-5GEA 461 EUR
APOSPC449018-5G
1-[3-(Trifluoromethyl)phenyl]-cyclobutanecarboxylic acid 98%
1-[3-(Trifluoromethyl)phenyl]-cyclobutanecarboxylic acid
Formel: C₁₂H₁₁F₃O₂
Molecular Weight: 244,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11037070
CAS-Nummer: 151157-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR