1-(3,4-Difluorophenyl)-cyclobutanecarboxylic acid

Lieferant: Apollo Scientific

PC449019-1G
APOSPC449019-1GEA 960 EUR
APOSPC449019-1G
1-(3,4-Difluorophenyl)-cyclobutanecarboxylic acid
1-(3,4-Difluorophenyl)-cyclobutanecarboxylic acid
Formel: C₁₁H₁₀F₂O₂
Molecular Weight: 212,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11037035
CAS-Nummer: 633317-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR