a-Cyclopropyl-4-fluoro-benzylamine

Lieferant: Apollo Scientific

PC449021-250MG
APOSPC449021-250MGEA 1160 EUR
APOSPC449021-250MG
a-Cyclopropyl-4-fluoro-benzylamine
a-Cyclopropyl-4-fluoro-benzylamine
Formel: C₁₀H₁₂FN
Lagertemperatur: Raumtemperatur
MDL: MFCD05191953
CAS-Nummer: 705-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR