3-(4-Trifluoromethylphenyl)pyrrolidine hydrochloride

Lieferant: Apollo Scientific

PC449023-1G
APOSPC449023-1GEA 1160 EUR
APOSPC449023-1G
3-(4-Trifluoromethylphenyl)pyrrolidine hydrochloride
3-(4-Trifluoromethylphenyl)pyrrolidine hydrochloride
Formel: C₁₁H₁₂F₃N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD03701254
CAS-Nummer: 1095545-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR