Cyclopentyl(4-fluorobenzyl)amine

Lieferant: Apollo Scientific

Synonyme: N-(4-Fluorobenzyl)cyclopentylamine

PC449024-1G
APOSPC449024-1GEA 873 EUR
APOSPC449024-1G
Cyclopentyl(4-fluorobenzyl)amine
Cyclopentyl(4-fluorobenzyl)amine
Formel: C₁₂H₁₆FN
Molecular Weight: 193,265 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03724763
CAS-Nummer: 85952-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR