4-[4-(Trifluoromethoxy)phenyl]-4-piperidinol hydrochloride

Lieferant: Apollo Scientific

PC449025-1G
APOSPC449025-1GEA 960 EUR
APOSPC449025-1G
4-[4-(Trifluoromethoxy)phenyl]-4-piperidinol hydrochloride
4-[4-(Trifluoromethoxy)phenyl]-4-piperidinol hydrochloride
Formel: C₁₂H₁₄F₃NO₂·ClH
Molecular Weight: 297,701 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28108997
CAS-Nummer: 871112-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR