4-(Trifluoromethoxy)indan-1-amine hydrochloride

Lieferant: Apollo Scientific

PC449026-500MG
APOSPC449026-500MGEA 1320 EUR
APOSPC449026-500MG
4-(Trifluoromethoxy)indan-1-amine hydrochloride
4-(Trifluoromethoxy)indan-1-amine hydrochloride
Formel: C₁₀H₁₀F₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28975214
CAS-Nummer: 1083090-92-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR