N-[3,5-Bis(trifluoromethyl)phenyl]-N'-[4-hydroxy-6-(trifluoromethyl)pyrimidin-2-yl)]guanidine

Lieferant: Apollo Scientific
Achtung

PC4493-1G
APOSPC4493-1GEA 140 EUR
APOSPC4493-1G
N-[3,5-Bis(trifluoromethyl)phenyl]-N'-[4-hydroxy-6-(trifluoromethyl)pyrimidin-2-yl)]guanidine
N-[3,5-Bis(trifluoromethyl)phenyl]-N'-[4-hydroxy-6-(trifluoromethyl)pyrimidin-2-yl)]guanidine
Formel: C₁₄H₈F₉N₅O
Molecular Weight: 433,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05150373
CAS-Nummer: 618092-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR