Methyl(2,2,2-trifluoroethyl)amine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC450006-1G PC450006-5G
APOSPC450006-1GEA 174 EUR
APOSPC450006-1G APOSPC450006-5G
Methyl(2,2,2-trifluoroethyl)amine hydrochloride
Methyl(2,2,2-trifluoroethyl)amine hydrochloride
Formel: C₃H₇ClF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD08448126
CAS-Nummer: 2730-52-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR