(R)-Boc-3-amino-4,4,4-trifluorobutyric acid

Lieferant: Apollo Scientific

PC450008-250MG
APOSPC450008-250MGEA 713 EUR
APOSPC450008-250MG
(R)-Boc-3-amino-4,4,4-trifluorobutyric acid
(R)-Boc-3-amino-4,4,4-trifluorobutyric acid
Formel: C₉H₁₄F₃NO₄
Molecular Weight: 257,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18762150
CAS-Nummer: 1310680-29-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR