(S)-Boc-3-amino-4,4,4-trifluorobutyric acid

Lieferant: Apollo Scientific

PC450009-250MG
APOSPC450009-250MGEA 388 EUR
APOSPC450009-250MG
(S)-Boc-3-amino-4,4,4-trifluorobutyric acid
(S)-Boc-3-amino-4,4,4-trifluorobutyric acid
Formel: C₉H₁₄F₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD18762152
CAS-Nummer: 1310680-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR