2-(2-Fluoro-5-methoxypyridin-3-yl)acetic acid

Lieferant: Apollo Scientific

PC450010-250MG
APOSPC450010-250MGEA 342 EUR
APOSPC450010-250MG
2-(2-Fluoro-5-methoxypyridin-3-yl)acetic acid
2-(2-Fluoro-5-methoxypyridin-3-yl)acetic acid
Formel: C₈H₈FNO₃
Molecular Weight: 185,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16611231
CAS-Nummer: 1227564-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR