(Z)-3-Fluoro-3-(2-methylphenyl)prop-2-enenitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(Z)-3-Fluoro-3-(2-methylphenyl)acrylonitrile
PC450012-100MG
APOSPC450012-100MGEA
285
EUR
APOSPC450012-100MG
(Z)-3-Fluoro-3-(2-methylphenyl)prop-2-enenitrile
(Z)-3-Fluoro-3-(2-methylphenyl)prop-2-enenitrile
Formel:
C₁₀H₈FN Molecular Weight: 161,176 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22124013 CAS-Nummer: 1365836-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...