3-(2-Iodoethyl)-4-(1H,1H-perfluoroheptyl)pyrrolidine-1-carbonitrile

Lieferant: Apollo Scientific

PC450013-250MG
APOSPC450013-250MGEA 575 EUR
APOSPC450013-250MG
3-(2-Iodoethyl)-4-(1H,1H-perfluoroheptyl)pyrrolidine-1-carbonitrile
3-(2-Iodoethyl)-4-(1H,1H-perfluoroheptyl)pyrrolidine-1-carbonitrile
Formel: C₁₄H₁₂F₁₃IN₂
Molecular Weight: 582,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22124040
CAS-Nummer: 1365808-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR