2,2-Difluoro-1-phenyl-2-(pyridin-3-yl)ethan-1-one

Lieferant: Apollo Scientific

PC450014-100MG
APOSPC450014-100MGEA 212 EUR
APOSPC450014-100MG
2,2-Difluoro-1-phenyl-2-(pyridin-3-yl)ethan-1-one
2,2-Difluoro-1-phenyl-2-(pyridin-3-yl)ethan-1-one
Formel: C₁₃H₉F₂NO
Molecular Weight: 233,213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22377945
CAS-Nummer: 1378261-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR