3-Brom-2,6-difluorphenol

Lieferant: Apollo Scientific
Achtung

PC450015-1G PC450015-5G
APOSPC450015-1GEA 85.7 EUR
APOSPC450015-1G APOSPC450015-5G
3-Brom-2,6-difluorphenol
3-Brom-2,6-difluorphenol
Formel: C₆H₃BrF₂O
Lagertemperatur: Kühlschrank
MDL: MFCD12910487
CAS-Nummer: 221220-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR