(S)-1,1,1,4,4,4-Hexafluoro-2-butylamine

Lieferant: Apollo Scientific

PC450016-250MG
APOSPC450016-250MGEA 569 EUR
APOSPC450016-250MG
(S)-1,1,1,4,4,4-Hexafluoro-2-butylamine
(S)-1,1,1,4,4,4-Hexafluoro-2-butylamine
Formel: C₄H₅F₆N
Molecular Weight: 181,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18917097
CAS-Nummer: 1349699-79-1
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR