2-[3-(Trifluoromethyl)phenyl]pyrrolidine

Lieferant: Apollo Scientific
Achtung

PC450017-250MG
APOSPC450017-250MGEA 136 EUR
APOSPC450017-250MG
2-[3-(Trifluoromethyl)phenyl]pyrrolidine
2-[3-(Trifluoromethyl)phenyl]pyrrolidine
Formel: C₁₁H₁₂F₃N
Lagertemperatur: Kühlschrank
MDL: MFCD02663523
CAS-Nummer: 109086-17-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR