2'-Fluoro-2'-deoxyguanosine

Lieferant: Apollo Scientific

PC450021-1G
APOSPC450021-1GEA 156 EUR
APOSPC450021-1G
2'-Fluoro-2'-deoxyguanosine
2'-Fluoro-2'-deoxyguanosine
Formel: C₁₀H₁₂FN₅O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00923832
CAS-Nummer: 78842-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR