Ethyl 2,2-difluoro-2-(3,4-dichlorophenyl)acetate

Lieferant: Apollo Scientific

PC450025-250MG PC450025-1G
APOSPC450025-250MGEA 139 EUR
APOSPC450025-250MG APOSPC450025-1G
Ethyl 2,2-difluoro-2-(3,4-dichlorophenyl)acetate
Ethyl 2,2-difluoro-2-(3,4-dichlorophenyl)acetate
Formel: C₁₀H₈Cl₂F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01937334
CAS-Nummer: 56177-76-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR