2-(3-Bromo-4-fluorophenyl)-4-(trifluoromethyl)-1H-imidazole
Lieferant: Apollo Scientific
PC450027-1G
APOSPC450027-1GEA
575
EUR
APOSPC450027-1G
2-(3-Bromo-4-fluorophenyl)-4-(trifluoromethyl)-1H-imidazole
2-(3-Bromo-4-fluorophenyl)-4-(trifluoromethyl)-1H-imidazole
Formel:
C₁₀H₅BrF₄N₂ Molecular Weight: 309,058 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442113 CAS-Nummer: 1309602-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...