(1S)-1-(2-Fluorophenyl)-N-[(1S)-1-phenylethyl]ethanaminium 2-carboxybenzoate

Lieferant: Apollo Scientific

PC450028-250MG
APOSPC450028-250MGEA 227 EUR
APOSPC450028-250MG
(1S)-1-(2-Fluorophenyl)-N-[(1S)-1-phenylethyl]ethanaminium 2-carboxybenzoate
(1S)-1-(2-Fluorophenyl)-N-[(1S)-1-phenylethyl]ethanaminium 2-carboxybenzoate
Formel: C₁₆H₁₉FN·C₈H₅O₄
Molecular Weight: 409,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28397824
CAS-Nummer: 1428344-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR