(1R)-1-Phenyl-N-{(1R)-1-[4-(trifluoromethyl)phenyl]ethyl}ethanaminium 2-carboxybenzoate
Lieferant: Apollo Scientific
PC450030-250MG
APOSPC450030-250MGEA
227
EUR
APOSPC450030-250MG
(1R)-1-Phenyl-N-{(1R)-1-[4-(trifluoromethyl)phenyl]ethyl}ethanaminium 2-carboxybenzoate
(1R)-1-Phenyl-N-{(1R)-1-[4-(trifluoromethyl)phenyl]ethyl}ethanaminium 2-carboxybenzoate
Formel:
C₁₇H₁₉F₃N·C₈H₅O₄ Molecular Weight: 459,458 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18642880 CAS-Nummer: 626244-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...