[(1S)-1-(3,5-Bis(Trifluoromethyl)phenyl)ethyl][(1S)-1-phenylethyl]ammonium p-tosylate

Lieferant: Apollo Scientific
Achtung

PC450031-250MG
APOSPC450031-250MGEA 227 EUR
APOSPC450031-250MG
[(1S)-1-(3,5-Bis(Trifluoromethyl)phenyl)ethyl][(1S)-1-phenylethyl]ammonium p-tosylate
[(1S)-1-(3,5-Bis(Trifluoromethyl)phenyl)ethyl][(1S)-1-phenylethyl]ammonium p-tosylate
Formel: C₁₈H₁₈F₆N·C₇H₇O₃S
Molecular Weight: 533,526 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28397826
CAS-Nummer: 384824-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR